Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033079.2 | NP_149070.2 | 7 | 1061 | |
GenBank mRNA | AF084364.1 | - | 6 | 1030 | |
AK131954.1 | - | 7 | 1061 | ||
BC119803.1 | - | 6 | 988 | 40107692 | |
BC122521.1 | - | 6 | 993 | 40107688 |