Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033074.3 | NP_149065.2 | 18 | 2037 | |
GenBank mRNA | AK010256.1 | - | 18 | 2004 | |
AK011146.2 | - | 18 | 2038 | ||
AK031064.1 | - | 18 | 2001 | ||
AK152945.1 | - | 18 | 1998 | ||
AK153314.1 | - | 18 | 2009 | ||
AK166693.1 | - | 18 | 2004 | ||
AK167597.1 | - | 18 | 2046 | ||
AK168969.1 | - | 18 | 1997 | ||
AK208469.1 | - | 3 | 197 | ||
AK209524.1 | - | 3 | 197 | ||
AK211015.1 | - | 3 | 197 | ||
AK213411.1 | - | 3 | 197 | ||
AK214483.1 | - | 3 | 191 | ||
AK215269.1 | - | 3 | 197 | ||
BC004621.1 | - | 11 | 1124 | ||
BC055371.1 | - | 18 | 2023 | 30060749 | |
BC060176.1 | - | 2132 |