Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007502.4 | NP_031528.1 | 2 | 363 | |
GenBank mRNA | AK032516.1 | - | 2 | 380 | |
AK151700.1 | - | 2 | 380 | ||
AK151841.1 | - | 2 | 379 | ||
AK159532.1 | - | 2 | 380 | ||
AK161282.1 | - | 2 | 395 | ||
AK168273.1 | - | 2 | 379 | ||
AK196817.1 | - | 2 | 144 | ||
BC079916.1 | - | 2 | 368 | 30665138 | |
U59761.1 | - | 2 | 366 |