Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009858.2 | NP_033988.1 | 2 | 137 | |
GenBank mRNA | AK087944.1 | - | 2 | 114 | |
AK088372.1 | - | 2 | 162 | ||
AK088649.1 | - | 2 | 161 | ||
AK154055.1 | - | 2 | 117 | ||
AK162993.1 | - | 2 | 126 | ||
AK169581.1 | - | 2 | 138 | ||
M16799.1 | - | 2 | 130 | ||
M17534.1 | - | 2 | 137 | ||
M19504.1 | - | 2 | 125 | ||
U34882.1 | - | 2 | 257 | ||
X07698.1 | - | 2 | 141 |