Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007819.4 | NP_031845.1 | 1, 2 | 166 | |
GenBank mRNA | AK136494.1 | - | 1, 2 | 165 | |
AK136529.1 | - | 1, 2 | 167 | ||
AK149572.1 | - | 1, 2 | 167 | ||
BC046592.1 | - | 1, 2 | 145 | 6308234 | |
X63023.1 | - | 1, 2 | 142 |