Gene Transcripts
- Gene Symbol
- Elmo2
- Entrez Gene ID
- 140579
- Gene Name
- engulfment and cell motility 2
- Gene Aliases
- 1190002F24Rik, CED-12
- Chromosome Location
- Chr. 2: 165288031 - 165326479 on Build GRCm38
- UniGene ID
- Mm.35064
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_080287.2 |
NP_525026.2
|
2, 3 |
163 |
|
NM_207706.1 |
NP_997589.1
|
2, 3 |
163 |
|
GenBank mRNA |
AF398884.1 |
- |
1, 2 |
68 |
|
AK032033.1 |
- |
2, 3 |
136 |
|
AK033918.1 |
- |
2, 3 |
357 |
|
AK038097.1 |
- |
2, 3 |
154 |
|
AK038455.1 |
- |
2, 3 |
269 |
|
AK045428.1 |
- |
2, 3 |
164 |
|
AK047040.1 |
- |
2, 3 |
125 |
|
AK050823.1 |
- |
2, 3 |
126 |
|
AK053574.1 |
- |
2, 3 |
127 |
|
AK129023.1 |
- |
2, 3 |
163 |
|
BC023954.1 |
- |
2, 3 |
148 |
5320429 |
BC033418.1 |
- |
1, 2 |
93 |
|
View Full Product Details