Gene Transcripts

Gene Symbol
Ganab
Entrez Gene ID
14376
Gene Name
alpha glucosidase 2 alpha neutral subunit
Gene Aliases
AU042638, G2an, GluII, mKIAA0088
Chromosome Location
Chr. 19: 8898057 - 8916742 on Build GRCm38
UniGene ID
Mm.3196
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293621.1 NP_001280550.1 17 2166
NM_008060.2 NP_032086.1 18 2232
GenBank mRNA AK017873.1 - 5 572
AK028655.1 - 17 2136
AK030722.1 - 17 2135
AK047454.1 - 9 2259
AK081915.1 - 17 2128
AK122201.1 - 12 2362
AK166150.1 - 17 2105
BC020386.1 - 10 1382
BC035423.1 - 8 982
BC094437.1 - 17 2116 30682580
BC117888.1 - 17 2109 40096413
BC117889.1 - 17 2112 40096414
BC128069.1 - 18 2176 40096428
U92793.1 - 18 2178

View Full Product Details