Gene Transcripts

Gene Symbol
Kcnh3
Entrez Gene ID
16512
Gene Name
potassium voltage-gated channel, subfamily H (eag-related), member 3
Gene Aliases
AU019351, C030044P22Rik, Elk2, Kv12.2, Melk2
Chromosome Location
Chr. 15: 99224358 - 99242826 on Build GRCm38
UniGene ID
Mm.374793
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010601.3 NP_034731.3 4, 5 590
GenBank mRNA AY380579.1 - 4, 5 590
BC141013.1 - 4, 5 705 9055625
BC145145.1 - 4, 5 705 9053696

View Full Product Details