Gene Transcripts

Gene Symbol
Oosp1
Entrez Gene ID
170834
Gene Name
oocyte secreted protein 1
Gene Aliases
AA536827
Chromosome Location
Chr. 19: 11667460 - 11691051 on Build GRCm38
UniGene ID
Mm.24754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133353.3 NP_579931.1 4, 5 515
GenBank mRNA AB050982.2 - 4, 5 508
AB086437.1 - 3, 4 400
AF420487.1 - 4, 5 520
AK033038.1 - 4, 5 518
AK162132.1 - 4, 5 518
BC100313.1 - 4, 5 494 30936071

View Full Product Details