Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010930.4 | NP_035060.1 | 4 | 737 | |
GenBank mRNA | AK039481.1 | - | 4 | 728 | |
AK081310.1 | - | 4 | 755 | ||
AK081944.1 | - | 4 | 963 | ||
AK136407.1 | - | 4 | 757 | ||
AK155048.1 | - | 4 | 756 | ||
BC003774.1 | - | 4 | 743 | 3494896 | |
X96585.1 | - | 4 | 737 | ||
Y09257.1 | - | 4 | 725 |