Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083316.1 | NP_001076785.1 | 8 | 1403 | |
NM_011058.2 | NP_035188.2 | 8 | 1386 | ||
GenBank mRNA | AK035501.1 | - | 8 | 1343 | |
AK081664.1 | - | 8 | 1385 | ||
AK136490.1 | - | 8 | 1372 | ||
AK139094.1 | - | 7 | 1618 | ||
AK147267.1 | - | 8 | 1335 | ||
AK163952.1 | - | 8 | 1450 | ||
BC020405.1 | - | 8 | 1317 | ||
BC037467.1 | - | 8 | 1325 | ||
BC053036.1 | - | 8 | 1403 | 5704645 | |
M57683.1 | - | 8 | 1285 | ||
M84607.1 | - | 8 | 1396 |