Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291019.1 | NP_001277948.1 | 3 | 1041 | |
NM_001291020.1 | NP_001277949.1 | 4 | 843 | ||
NM_007547.4 | NP_031573.2 | 3 | 1041 | ||
GenBank mRNA | AB018194.1 | - | 3 | 607 | |
AF072543.1 | - | 3 | 597 | ||
AF332079.1 | - | 3 | 607 | ||
AF332080.1 | - | 3 | 607 | ||
AK159617.1 | - | 4 | 848 | ||
AK193777.1 | - | 1 | 63 | ||
AK197651.1 | - | 1 | 63 | ||
AK201986.1 | - | 63 | |||
BC062197.1 | - | 3 | 632 | 5368250 | |
D85785.1 | - | 3 | 891 | ||
D87967.1 | - | 3 | 1006 | ||
D87968.1 | - | 3 | 1006 | ||
U89694.1 | - | 3 | 618 | ||
Y10349.1 | - | 3 | 652 |