Gene Transcripts

Gene Symbol
Arhgdia
Entrez Gene ID
192662
Gene Name
Rho GDP dissociation inhibitor (GDI) alpha
Gene Aliases
5330430M07Rik, Gdi-1, Rho-GDI, RhoDGI, RhoGDI-1
Chromosome Location
Chr. 11: 120577235 - 120581620 on Build GRCm38
UniGene ID
Mm.30016
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133796.7 NP_598557.3 4 413
GenBank mRNA AK075656.1 - 3 280
AK077343.1 - 4 573
AK143516.1 - 4 397
AK147842.1 - 372
AK156819.1 - 358
AK165241.1 - 4 395
AK170553.1 - 4 380
AK170718.1 - 4 397
AK172140.1 - 358
AK204953.1 - 2 95
BC004732.1 - 4 355 3583026
BC086755.1 - 4 337 6414768

View Full Product Details