Gene Transcripts

Gene Symbol
Rad17
Entrez Gene ID
19356
Gene Name
RAD17 homolog (S. pombe)
Gene Aliases
MmRad24
Chromosome Location
Chr. 13: 100617164 - 100651061 on Build GRCm38
UniGene ID
Mm.248489
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044371.2 NP_001037836.1 6 824
NM_001283011.1 NP_001269940.1 7 1029
NM_011233.3 NP_035363.2 6 952
GenBank mRNA AF085737.1 - 6 875
AJ011923.1 - 5 725
AK134277.1 - 6 779
AK162016.1 - 6 811
AK167358.1 - 7 972
BC023939.1 - 6 829

View Full Product Details