Gene Transcripts

Gene Symbol
Slc12a7
Entrez Gene ID
20499
Gene Name
solute carrier family 12, member 7
Gene Aliases
AA408796, D13Ertd261e, Kcc4
Chromosome Location
Chr. 13: 73733053 - 73816755 on Build GRCm38
UniGene ID
Mm.275800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011390.2 NP_035520.1 8 1018
GenBank mRNA AF087436.1 - 8 1020
AK052910.1 - 8 1069
AK083598.1 - 5 3289
AK131129.1 - 8 972
AK143535.1 - 8 1012
AK149750.1 - 8 1013
AK166215.1 - 8 1012
AK169950.1 - 8 1008
AK171498.1 - 8 1055
BC059242.1 - 8 1010
BC060133.1 - 8 1010
BC079677.1 - 8 976
BC141107.1 - 8 978 9055719

View Full Product Details