Gene Transcripts
- Gene Symbol
- Stau1
- Entrez Gene ID
- 20853
- Gene Name
- staufen (RNA binding protein) homolog 1 (Drosophila)
- Gene Aliases
- 5830401L18Rik, AW549911, C85792, Stau
- Chromosome Location
- Chr. 2: 166947549 - 166996299 on Build GRCm38
- UniGene ID
- Mm.73276
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001109905.2 |
NP_001103375.1
|
9, 10 |
1567 |
|
NM_001109906.2 |
NP_001103376.1
|
9, 10 |
1585 |
|
NM_001291160.1 |
NP_001278089.1
|
9, 10 |
1567 |
|
NM_011490.4 |
NP_035620.1
|
9, 10 |
1561 |
|
GenBank mRNA |
AF061942.1 |
- |
9, 10 |
1561 |
|
AF395842.1 |
- |
8, 9 |
1288 |
|
AK005002.2 |
- |
9, 10 |
1508 |
|
AK139532.1 |
- |
9, 10 |
1514 |
|
AK145826.1 |
- |
9, 10 |
1552 |
|
AK151835.1 |
- |
9, 10 |
1373 |
|
AK152269.1 |
- |
9, 10 |
1379 |
|
AK152744.1 |
- |
9, 10 |
1373 |
|
AK158952.1 |
- |
4, 5 |
2148 |
|
AK159602.1 |
- |
9, 10 |
1502 |
|
AK160979.1 |
- |
9, 10 |
1526 |
|
AK167637.1 |
- |
9, 10 |
1508 |
|
AK179726.1 |
- |
1, 2 |
67 |
|
AY391773.1 |
- |
9, 10 |
1233 |
|
BC012959.1 |
- |
9, 10 |
1531 |
4188394 |
BC082277.1 |
- |
9, 10 |
1504 |
|
View Full Product Details