Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_199364.2 | NP_955396.2 | 3 | 1189 | |
GenBank mRNA | AF288601.1 | - | 3 | 1189 | |
BC127606.1 | - | 3 | 1188 | ||
BC127607.1 | - | 3 | 1189 | 40057969 | |
BC139350.1 | - | 3 | 1245 | 8862372 | |
BC139352.1 | - | 3 | 1245 | 8862374 |