Gene Transcripts

Gene Symbol
Cpne9
Entrez Gene ID
211232
Gene Name
copine family member IX
Gene Aliases
A730016F12Rik, mKIAA4217
Chromosome Location
Chr. 6: 113282266 - 113305571 on Build GRCm38
UniGene ID
Mm.65597
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170673.3 NP_733773.2 17 1321
GenBank mRNA AK042692.1 - 17 1322
AK044080.1 - 17 1287
AK047571.1 - 15 2336
AK137422.1 - 17 1322
AK220550.1 - 9 2685

View Full Product Details