Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013414.3 | NP_001013432.1 | 10 | 1940 | |
NM_001291118.1 | NP_001278047.1 | 10 | 1940 | ||
NM_001291119.1 | NP_001278048.1 | 9 | 1874 | ||
GenBank mRNA | AK157107.1 | - | 9 | 1740 | |
AK173264.1 | - | 9 | 1435 |