Gene Transcripts

Gene Symbol
G2e3
Entrez Gene ID
217558
Gene Name
G2/M-phase specific E3 ubiquitin ligase
Gene Aliases
6030408C04Rik, 9030416F18, AW046379, D930034K21Rik, mKIAA1333
Chromosome Location
Chr. 12: 51348230 - 51376986 on Build GRCm38
UniGene ID
Mm.253264
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015099.2 NP_001015099.2 9 1069
NM_001167963.1 NP_001161435.1 9 1069
NM_001167964.1 NP_001161436.1 9 1069
GenBank mRNA AK031333.1 - 9 929
AK033513.1 - 9 1011
AK084256.1 - 9 940
AK129332.1 - 9 972
BC086455.1 - 9 942 30458160

View Full Product Details