Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009373.3 | NP_033399.1 | 5 | 677 | |
GenBank mRNA | AF076928.1 | - | 4 | 592 | |
AK052912.1 | - | 5 | 652 | ||
AK054108.1 | - | 5 | 652 | ||
AK080224.1 | - | 5 | 677 | ||
AK080593.1 | - | 5 | 652 | ||
AK086001.1 | - | 5 | 652 | ||
AK089481.1 | - | 5 | 680 | ||
AK143712.1 | - | 5 | 713 | ||
AK151776.1 | - | 5 | 678 | ||
AK152152.1 | - | 5 | 652 | ||
AK152627.1 | - | 5 | 678 | ||
AK159255.1 | - | 5 | 652 | ||
AK166302.1 | - | 5 | 652 | ||
AK168990.1 | - | 5 | 653 | ||
AK169356.1 | - | 5 | 680 | ||
BC016492.1 | - | 5 | 618 | 3256943 |