Gene Transcripts

Gene Symbol
Zfp13
Entrez Gene ID
22654
Gene Name
zinc finger protein 13
Gene Aliases
4933429B21, AI835008, Krox-8, Rhit, Zfp-13
Chromosome Location
Chr. 17: 23575844 - 23599516 on Build GRCm38
UniGene ID
Mm.40326
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011747.2 NP_035877.1 2, 3 469
GenBank mRNA AK030240.1 - 2, 3 470
BC052082.1 - 2, 3 372 6404424
FJ593171.1 - 2, 3 405

View Full Product Details