Gene Transcripts

Gene Symbol
Bahd1
Entrez Gene ID
228536
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
AL022997, AW541238, Gm117, mKIAA0945
Chromosome Location
Chr. 2: 118900332 - 118924528 on Build GRCm38
UniGene ID
Mm.330487
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045523.1 NP_001038988.1 5 2122
GenBank mRNA AK129243.1 - 5 2280
AK152597.1 - 5 2061
BC047433.1 - 4 871
BC100358.1 - 5 2284 30848279
BC141046.1 - 4 1985 9055658
BC145233.1 - 4 1985 9053785

View Full Product Details