Gene Transcripts

Gene Symbol
Vmn2r54
Entrez Gene ID
666085
Gene Name
vomeronasal 2, receptor 54
Gene Aliases
EG666085, Gm1496, Gm470
Chromosome Location
Chr. 7: 12615233 - 12636134 on Build GRCm38
UniGene ID
Mm.389810
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081449.2 NP_001074918.2 1 264

View Full Product Details