Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181517.3 | NP_852658.2 | 20, 21 | 2266 | |
GenBank mRNA | AK039534.1 | - | 20, 21 | 2418 | |
AK203124.1 | - | 13 | |||
AY316153.1 | - | 20, 21 | 2266 | ||
BC053524.1 | - | 17, 18 | 1949 | ||
BC064825.1 | - | 11, 12 | 1222 |