Gene Transcripts

Gene Symbol
Trim60
Entrez Gene ID
234329
Gene Name
tripartite motif-containing 60
Gene Aliases
2czf45, Rnf33
Chromosome Location
Chr. 8: 64998448 - 65018688 on Build GRCm38
UniGene ID
Mm.28010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153097.2 NP_694737.1 2 921
GenBank mRNA AF290197.1 - 2 823
AK135841.1 - 2 818
BC139468.1 - 2 805 8862490
BC139469.1 - 2 805 8862491

View Full Product Details