Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177546.2 | NP_808214.1 | 4 | 694 | |
NM_211138.1 | NP_997593.1 | 4 | 705 | ||
GenBank mRNA | AK032027.1 | - | 4 | 407 | |
AK053577.1 | - | 4 | 679 | ||
AK149304.1 | - | 4 | 695 | ||
AY189949.1 | - | 4 | 542 | ||
AY189950.1 | - | 4 | 694 | ||
BC048917.1 | - | 4 | 576 | 6309383 |