Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022813.3 | NP_073724.1 | 5, 6 | 587 | |
GenBank mRNA | AF295402.1 | - | 5, 6 | 591 | |
AK003080.1 | - | 5, 6 | 589 | ||
AK161002.1 | - | 5, 6 | 587 | ||
AK171766.1 | - | 5, 6 | 578 | ||
AK186279.1 | - | 1, 2 | 39 | ||
BC014751.1 | - | 5, 6 | 564 | 4235556 |