Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081315.1 | NP_001074784.1 | 8 | 2943 | |
GenBank mRNA | AK129322.1 | - | 6 | 1198 | |
AK147555.1 | - | 8 | 2942 | ||
BC157915.1 | - | 7 | 2708 | 9088158 | |
BC157957.1 | - | 7 | 2708 | ||
BC172121.1 | - | 7 | 2723 |