Gene Transcripts

Gene Symbol
Colgalt2
Entrez Gene ID
269132
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
AI427652, D730024P12, Glt25d2
Chromosome Location
Chr. 1: 152399867 - 152510695 on Build GRCm38
UniGene ID
Mm.23782
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177756.4 NP_808424.3 10, 11 1666
GenBank mRNA AK052838.1 - 10, 11 1667
AK076222.1 - 10, 11 1409
AK134084.1 - 10, 11 1647
BC068118.1 - 10, 11 1612 5706069

View Full Product Details