Gene Transcripts

Gene Symbol
Gnl3
Entrez Gene ID
30877
Gene Name
guanine nucleotide binding protein-like 3 (nucleolar)
Gene Aliases
Ns
Chromosome Location
Chr. 14: 31012433 - 31019131 on Build GRCm38
UniGene ID
Mm.88512
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153547.6 NP_705775.2 7 796
GenBank mRNA AK034825.1 - 1 1614
AK077523.1 - 7 727
AK167094.1 - 5 2229
AK167183.1 - 7 680
AY181025.1 - 7 654
AY185498.1 - 7 654
BC018560.1 - 7 635
BC037996.1 - 7 656 5052858

View Full Product Details