Gene Transcripts

Gene Symbol
Svopl
Entrez Gene ID
320590
Gene Name
SV2 related protein homolog (rat)-like
Gene Aliases
9430071P14Rik
Chromosome Location
Chr. 6: 37983739 - 38047014 on Build GRCm38
UniGene ID
Mm.197449
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177200.4 NP_796174.2 6, 7 575
GenBank mRNA AK034997.1 - 6, 7 576
AK137043.1 - 6, 7 576
BB627230.1 - 6, 7 576
BC058741.1 - 6, 7 558 6309979

View Full Product Details