Gene Transcripts

Gene Symbol
Mslnl
Entrez Gene ID
328783
Gene Name
mesothelin-like
Gene Aliases
4732467B22
Chromosome Location
Chr. 17: 25733503 - 25748530 on Build GRCm38
UniGene ID
Mm.183275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177822.3 NP_808490.1 15, 16 2192
GenBank mRNA AK028891.1 - 15, 16 2192
BC052484.1 - 14, 15 1928 6312790

View Full Product Details