Gene Transcripts

Gene Symbol
Rbm44
Entrez Gene ID
329207
Gene Name
RNA binding motif protein 44
Gene Aliases
Gm817
Chromosome Location
Chr. 1: 91144973 - 91170799 on Build GRCm38
UniGene ID
Mm.295940
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033408.4 NP_001028580.1 14 2968
GenBank mRNA AK133355.1 - 14 2969
BC150950.1 - 14 2930 9087861

View Full Product Details