Gene Transcripts

Gene Symbol
Mia3
Entrez Gene ID
338366
Gene Name
melanoma inhibitory activity 3
Gene Aliases
9130229H14Rik, A930039G15Rik, AU022327, B230399H06Rik, C80126, Gm1525, TANGO1, Tango
Chromosome Location
Chr. 1: 183326236 - 183369565 on Build GRCm38
UniGene ID
Mm.41152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177389.3 NP_796363.2 4 2793
GenBank mRNA AK044749.1 - 4 2770

View Full Product Details