Gene Transcripts
- Gene Symbol
- Gars
- Entrez Gene ID
- 353172
- Gene Name
- glycyl-tRNA synthetase
- Gene Aliases
- GENA202, Gena201, Nmf249, Sgrp23
- Chromosome Location
- Chr. 6: 55038001 - 55079504 on Build GRCm38
- UniGene ID
- Mm.250004
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_180678.3 |
NP_851009.2
|
8, 9 |
1036 |
|
GenBank mRNA |
AK012758.2 |
- |
8, 9 |
1031 |
|
AK146238.1 |
- |
8, 9 |
1013 |
|
AK165857.1 |
- |
8, 9 |
984 |
|
AK190416.1 |
- |
1, 2 |
100 |
|
AK190448.1 |
- |
2, 3 |
202 |
|
AK191539.1 |
- |
2, 3 |
223 |
|
AK194283.1 |
- |
2, 3 |
178 |
|
AK195104.1 |
- |
2, 3 |
183 |
|
AK196051.1 |
- |
2, 3 |
202 |
|
BC021747.1 |
- |
8, 9 |
1017 |
4192628 |
View Full Product Details