Gene Transcripts

Gene Symbol
Gars
Entrez Gene ID
353172
Gene Name
glycyl-tRNA synthetase
Gene Aliases
GENA202, Gena201, Nmf249, Sgrp23
Chromosome Location
Chr. 6: 55038001 - 55079504 on Build GRCm38
UniGene ID
Mm.250004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_180678.3 NP_851009.2 7 870
GenBank mRNA AK012758.2 - 7 865
AK146238.1 - 7 847
AK165857.1 - 7 818
AK183945.1 - 3 246
AK190448.1 - 1 36
AK191539.1 - 1 57
AK194283.1 - 1 12
AK195104.1 - 1 17
AK196051.1 - 1 36
AK212952.1 - 1 36
BC021747.1 - 7 851 4192628

View Full Product Details