Gene Transcripts

Gene Symbol
Cpxcr1
Entrez Gene ID
382239
Gene Name
CPX chromosome region, candidate 1
Gene Aliases
Gm1143
Chromosome Location
Chr. X: 116448944 - 116478783 on Build GRCm38
UniGene ID
Mm.336224
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033471.3 NP_001028643.2 3 326
GenBank mRNA AK133001.1 - 3 328
BC147520.1 - 2 183 9056512
BC147548.1 - 2 183 9056539

View Full Product Details