Gene Transcripts

Gene Symbol
Kbtbd6
Entrez Gene ID
432879
Gene Name
kelch repeat and BTB (POZ) domain containing 6
Gene Aliases
EG432879, Gm5465, Zbtbd6
Chromosome Location
Chr. 14: 79451835 - 79454816 on Build GRCm38
UniGene ID
Mm.308666
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034882.3 NP_001030054.3 1 1886
GenBank mRNA AK159684.1 - 1 1887
AK163295.1 - 2 1842

View Full Product Details