Gene Transcripts

Gene Symbol
Spast
Entrez Gene ID
50850
Gene Name
spastin
Gene Aliases
Spg4, mKIAA1083
Chromosome Location
Chr. 17: 74337454 - 74391115 on Build GRCm38
UniGene ID
Mm.19804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162870.1 NP_001156342.1 6 1007
NM_016962.2 NP_058658.2 6 1004
GenBank mRNA AJ246002.1 - 6 629
AK007793.1 - 6 784
AK044900.1 - 6 859
AK129282.1 - 6 981
BC046286.1 - 6 1004 6441742

View Full Product Details