Gene Transcripts

Gene Symbol
Pdgfc
Entrez Gene ID
54635
Gene Name
platelet-derived growth factor, C polypeptide
Gene Aliases
1110064L01Rik, AI647969, PDGF-C
Chromosome Location
Chr. 3: 81035885 - 81214040 on Build GRCm38
UniGene ID
Mm.331089
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019971.2 NP_064355.1 3 1491
GenBank mRNA AF117608.1 - 3 667
AF266467.1 - 3 1491
AF286725.1 - 3 470
AK033734.1 - 3 1220
AK042767.1 - 3 1238
AK052947.1 - 3 743
AK081347.1 - 4 543
BC029099.1 - 4 794
BC037696.1 - 3 678 4008749

View Full Product Details