Gene Transcripts

Gene Symbol
Crtam
Entrez Gene ID
54698
Gene Name
cytotoxic and regulatory T cell molecule
Gene Aliases
Chromosome Location
Chr. 9: 40972798 - 41004628 on Build GRCm38
UniGene ID
Mm.42791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281954.1 NP_001268883.1 9 1043
NM_019465.4 NP_062338.3 9 1040
GenBank mRNA AF001104.1 - 9 986
AK163744.1 - 4 441
BC117715.1 - 9 988
BC117716.1 - 9 989 40088099

View Full Product Details