Gene Transcripts

Gene Symbol
Tdp2
Entrez Gene ID
56196
Gene Name
tyrosyl-DNA phosphodiesterase 2
Gene Aliases
D13Ertd656e, Ttrap
Chromosome Location
Chr. 13: 24831647 - 24842153 on Build GRCm38
UniGene ID
Mm.427162
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019551.2 NP_062424.1 3 432
GenBank mRNA AJ251328.1 - 3 406
AK009089.1 - 3 409
AK135925.1 - 3 433
AK167707.1 - 2 228
BC132511.1 - 3 395 40130788
BC132513.1 - 3 395 40130790
BY719852.1 - 3 432

View Full Product Details