Gene Transcripts
- Gene Symbol
- Rgr
- Entrez Gene ID
- 57811
- Gene Name
- retinal G protein coupled receptor
- Gene Aliases
- Chromosome Location
- Chr. 14: 37034909 - 37049014 on Build GRCm38
- UniGene ID
- Mm.20460
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301692.1 |
NP_001288621.1
|
4, 5 |
596 |
|
NM_001301694.1 |
NP_001288623.1
|
4, 5 |
546 |
|
NM_021340.4 |
NP_067315.1
|
5, 6 |
718 |
|
GenBank mRNA |
AF076930.1 |
- |
5, 6 |
638 |
|
AK044450.1 |
- |
5, 6 |
670 |
|
AK044649.1 |
- |
5, 6 |
669 |
|
AK149272.1 |
- |
5, 6 |
669 |
|
BC082611.1 |
- |
5, 6 |
655 |
|
BC086689.1 |
- |
5, 6 |
645 |
|
BC119055.1 |
- |
5, 6 |
652 |
8733804 |
BC144937.1 |
- |
4, 5 |
531 |
|
CK623457.1 |
- |
4, 5 |
477 |
|
CK626179.1 |
- |
4, 5 |
534 |
|
View Full Product Details