Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021429.3 | NP_067404.2 | 2 | 153 | |
GenBank mRNA | AJ132192.1 | - | 2 | 147 | |
AK040654.1 | - | 2 | 153 | ||
AK084802.1 | - | 2 | 146 | ||
AK154696.1 | - | 2 | 149 | ||
AK170838.1 | - | 2 | 140 | ||
BC095977.1 | - | 2 | 150 |