Gene Transcripts

Gene Symbol
Lpin3
Entrez Gene ID
64899
Gene Name
lipin 3
Gene Aliases
9130206L11Rik, AA438110, AV236139, mKIAA4023
Chromosome Location
Chr. 2: 160880670 - 160906000 on Build GRCm38
UniGene ID
Mm.292111
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199118.1 NP_001186047.1 5, 6 878
NM_022883.3 NP_075021.1 5, 6 955
GenBank mRNA AF286724.1 - 5, 6 892
AK033647.1 - 5, 6 876
AK049363.1 - 5, 6 871
AK163833.1 - 5, 6 870
AK220228.1 - 5, 6 955
BC026729.1 - 5, 6 862
BC117882.1 - 4, 5 819 40096038
BC117883.1 - 4, 5 819 40096041

View Full Product Details