Gene Transcripts

Gene Symbol
Asb3
Entrez Gene ID
65257
Gene Name
ankyrin repeat and SOCS box-containing 3
Gene Aliases
2400011J03Rik, Asb-3
Chromosome Location
Chr. 11: 30954194 - 31102704 on Build GRCm38
UniGene ID
Mm.440539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023906.3 NP_076395.2 10 1624
GenBank mRNA AF155354.1 - 9 1439
AK010315.1 - 10 1560
AK028489.1 - 11 1693
AK143580.1 - 10 1559
BC023086.1 - 10 1558 4947610

View Full Product Details