Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163536.1 | NP_001157008.1 | 2 | 208 | |
NM_001163537.1 | NP_001157009.1 | 2 | 228 | ||
NM_025489.3 | NP_079765.3 | 2 | 215 | ||
GenBank mRNA | AA426920.1 | - | 2 | 282 | |
AK005725.1 | - | 2 | 195 | ||
AK005749.1 | - | 2 | 184 | ||
BC049579.1 | - | 2 | 158 | 6772915 | |
BY085815.1 | - | 2 | 209 |