Gene Transcripts

Gene Symbol
Ube2t
Entrez Gene ID
67196
Gene Name
ubiquitin-conjugating enzyme E2T (putative)
Gene Aliases
2700084L22Rik, C80607
Chromosome Location
Chr. 1: 134962565 - 134974180 on Build GRCm38
UniGene ID
Mm.284587
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278115.1 NP_001265044.1 2, 3 224
NM_026024.3 NP_080300.1 2, 3 401
GenBank mRNA AK012565.1 - 2, 3 360
AK021213.1 - 2, 3 390
AK166098.1 - 2, 3 371
BC029213.1 - 2, 3 350 3591557
CK334733.1 - 2, 3 215

View Full Product Details