Gene Transcripts

Gene Symbol
Ctdp1
Entrez Gene ID
67655
Gene Name
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) phosphatase, subunit 1
Gene Aliases
4930563P03Rik, AW553592
Chromosome Location
Chr. 18: 80407959 - 80470612 on Build GRCm38
UniGene ID
Mm.312893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026295.2 NP_080571.2 10 2406
GenBank mRNA AK156617.1 - 10 2435
BC053435.1 - 10 2406 6394578

View Full Product Details